Learn More
Biomatik Corporation High Sensitive Rat N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10310
Encompass_Preferred
The High Sensitive Rat N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit allows for quantitative determination of N-Terminal Pro-Brain Natriuretic Peptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 24.69-2000 pg/mL. Sensitivity: 9.15 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P13205. Cross Activity: No significant cross-reactivity or interference between N-Terminal Pro-Brain Natriuretic Peptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4518