Learn More
Biomatik Corporation High Sensitive Human Prothrombin Fragment 1+2 (F1+2) ELISA Kit, 96 tests
Catalog No. 501504519
Encompass_Preferred
The High Sensitive Human Prothrombin Fragment 1+2 (F1+2) ELISA Kit allows for quantitative determination of Prothrombin Fragment 1+2 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.11-80 pg/mL. Sensitivity: 0.04 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00734. Cross Activity: No significant cross-reactivity or interference between Prothrombin Fragment 1+2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4519 Supplier Biomatik Corporation Supplier No. EKU10314
The High Sensitive Human Prothrombin Fragment 1+2 (F1+2) ELISA Kit allows for quantitative determination of Prothrombin Fragment 1+2 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.11-80 pg/mL. Sensitivity: 0.04 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00734. Cross Activity: No significant cross-reactivity or interference between Prothrombin Fragment 1+2 and analogues was observed. Restriction: For Research Use Only.