Learn More
Biomatik Corporation Guanosine (GS) ELISA Kit
Supplier:  Biomatik Corporation EKU10886
Encompass_Preferred
Guanosine (GS) ELISA Kit for detecting GS in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 61.7-5,000pg/ml. Sensitivity: 26.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Guanine Riboside. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6790