Learn More
Biomatik Corporation Human Elastin (ELN) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10923
Encompass_Preferred
Human Elastin (ELN) High-sensitive ELISA Kit for detecting ELN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.47-30ng/ml. Sensitivity: 0.19ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15502. Gene ID: 2006. Target Synonyms: WBS, WS, SVAS; Tropoelastin; Supravalvular Aortic Stenosis; Williams-Beuren Syndrome. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6801