Learn More
Biomatik Corporation Wide-range Human Gelsolin (GS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10550
Encompass_Preferred
Human Gelsolin (GSN) Wide-range ELISA Kit for detecting GSN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 2.77ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06396. Gene ID: 2934. Target Synonyms: GS; AGEL; ADF; Brevin; Gelsolin Amyloidosis,Finnish Type; Actin-depolymerizing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4524