Learn More
Biomatik Corporation Wide-range Human Low Density Lipoprotein Receptor Related Protein 2 (LRP2) ELISA Kit, 96 tests
Catalog No. 501504525
Encompass_Preferred
Human Low Density Lipoprotein Receptor Related Protein 2 (LRP2) Wide-range ELISA Kit for detecting LRP2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P98164. Gene ID: 4036. Target Synonyms: DBS; gp330; Glycoprotein 330; Megalin; Low Density Lipoprotein-Related Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4525 Supplier Biomatik Corporation Supplier No. EKU10560
Human Low Density Lipoprotein Receptor Related Protein 2 (LRP2) Wide-range ELISA Kit for detecting LRP2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P98164. Gene ID: 4036. Target Synonyms: DBS; gp330; Glycoprotein 330; Megalin; Low Density Lipoprotein-Related Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.