Learn More
Biomatik Corporation Wide-range Human Programmed Cell Death Protein 1 Ligand 1 (PDL1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU10570
Encompass_Preferred
Human Programmed Cell Death Protein 1 Ligand 1 (PDL1) Wide-range ELISA Kit for detecting PDL1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NZQ7. Gene ID: 29126. Target Synonyms: CD274; PDCD1LG1; B7-H; B7H1; PD-L1; PDCD1L1; PDCD1-LG1; Programed Death Ligand 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4527