Learn More
Biomatik Corporation Human Receptor Interacting Serine Threonine Kinase 1 (RIPK1) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10959
Encompass_Preferred
Human Receptor Interacting Serine Threonine Kinase 1 (RIPK1) High-sensitive ELISA Kit for detecting RIPK1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13546. Gene ID: 8737. Target Synonyms: RIP; Receptor-Interacting Protein; Cell death protein RIP; Serine/threonine-protein kinase RIP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6819