Learn More
Biomatik Corporation Human F-actin ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC33652
Encompass_Preferred
The Human F-actin ELISA Kit allows for quantitative determination of F-actin in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-7885