Learn More
Biomatik Corporation Human Fibroblast growth factor 20 (FGF20) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC33695
Encompass_Preferred
The Human Fibroblast growth factor 20 (FGF20) ELISA Kit allows for quantitative determination of fibroblast growth factor 20 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 12.5 pg/mL-800 pg/mL. Sensitivity: 3.12 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9NP95. Restriction: For Research Use Only.
Catalog No. 50-149-7898