Learn More
Biomatik Corporation Human secreted frizzled-related protein 4 (SFRP4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35464
Encompass_Preferred
Human Secreted Frizzled-Related Protein 4 (Sfrp4) Elisa Kit for detecting Secreted Frizzled-Related Protein 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6FHJ7; SFRP4. Target Synonyms: Frizzled protein; Frizzled protein human endometrium; FRP 4; FRP4; FrpHE; human endometrium; PYL; Secreted frizzled-related protein 4; sFRP-4; Sfrp4; SFRP4_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8251