Learn More
Biomatik Corporation Human thioredoxin reductase, TrxR ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35759
Encompass_Preferred
Human Thioredoxin Reductase, Trxr Elisa Kit for detecting Thioredoxin Reductase 1 in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q16881; TXNRD1. Target Synonyms: cytoplasmic; Gene associated with retinoic and IFN-induced mortality 12 protein; Gene associated with retinoic and interferon-induced mortality 12 protein; Gene associated with retinoid IFN induced mortality 12 protein; GRIM 12; GRIM-12; GRIM12; KDRF; KM 102 derived reductase like factor; KM-102-derived reductase-like factor; MGC9145; Oxidoreductase; Thioredoxin reductase 1; Thioredoxin reductase 1 cytoplasmic; Thioredoxin reductase GRIM 12; Thioredoxin reductase TR1; TR 1; TR; TR1; TRXR 1; TRXR1; TRXR1_HUMAN; TXNR; TXNRD 1; Txnrd1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8306