Learn More
Biomatik Corporation Human Thioredoxin, Trx ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35761
Encompass_Preferred
Human Thioredoxin, Trx Elisa Kit for detecting Thioredoxin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 4.688-300ng/ml. Sensitivity: 1.172ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10599; TXN. Target Synonyms: ADF; ATL derived factor; ATL-derived factor; DKFZp686B1993; MGC61975; SASP; Surface associated sulphydryl protein; Surface-associated sulphydryl protein; testicular tissue protein Li 199; THIO_HUMAN; Thioredoxin; thioredoxin delta 3; TRDX; TRX 1; Trx; TRX1; TXN; TXN delta 3; TXN protein; zgc:92903. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8307