Learn More
Biomatik Corporation Mouse Interferon Gamma (IFNg) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11152
Encompass_Preferred
Mouse Interferon Gamma (IFNg) Mini sample ELISA Kit for detecting IFNg in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01580. Gene ID: 15978. Target Synonyms: IFN-G; IFG; IFI; INFr; IFN, Immune Interferon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6858