Learn More
Biomatik Corporation Mouse Protein argonaute-2 (EIF2C2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37649
Encompass_Preferred
Mouse Protein Argonaute-2 (Eif2C2) Elisa Kit for detecting Eukaryotic Translation Initiation Factor 2C, 2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8CJG0; Ago2. Target Synonyms: Ago2; Eif2c2; Kiaa4215; Protein argonaute-2; Argonaute2; mAgo2; EC 3.1.26.n2; Argonaute RISC catalytic component 2; Eukaryotic translation initiation factor 2C 2; eIF-2C 2; eIF2C 2; Piwi/argonaute family protein meIF2C2; Protein slicer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8648