Learn More
Biomatik Corporation Mouse Triggering receptor expressed on myeloid cells 2 (TREM2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37962
Encompass_Preferred
Mouse Triggering Receptor Expressed On Myeloid Cells 2 (Trem2) Elisa Kit for detecting Triggering Receptor Expressed On Myeloid Cells 2 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q99NH8; Trem2. Target Synonyms: Trem2; Trem2a; Trem2b; Trem2c; Triggering receptor expressed on myeloid cells 2; TREM-2; Triggering receptor expressed on monocytes 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8701