Learn More
Biomatik Corporation Pig calprotectin (CALP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38116
Encompass_Preferred
Pig Calprotectin (Calp) Elisa Kit for detecting Calprotectin in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8725