Learn More
Biomatik Corporation Human Apolipoprotein B48 (APOB48) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02506
Encompass_Preferred
The Human Apolipoprotein B48 (APOB48) ELISA Kit allows for quantitative determination of Apolipoprotein B48 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.47-30 ng/mL. Sensitivity: 0.17 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04114. Cross Activity: No significant cross-reactivity or interference between Apolipoprotein B48 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1849