Learn More
Biomatik Corporation Human Ferroportin (FPN) ELISA Kit, 96 tests
Catalog No. 501502470
Encompass_Preferred
The Human Ferroportin (FPN) ELISA Kit allows for quantitative determination of Ferroportin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.116 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NP59. Cross Activity: No significant cross-reactivity or interference between Ferroportin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2470 Supplier Biomatik Corporation Supplier No. EKU04079
The Human Ferroportin (FPN) ELISA Kit allows for quantitative determination of Ferroportin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.116 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NP59. Cross Activity: No significant cross-reactivity or interference between Ferroportin and analogues was observed. Restriction: For Research Use Only.