Learn More
Biomatik Corporation Mouse Heat Shock Protein 90kDa Alpha B1 (HSP90aB1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04678
Encompass_Preferred
Mouse Heat Shock Protein 90kDa Alpha B1 (HSP90aB1) ELISA Kit for detecting HSP90aB1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.56ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11499. Gene ID: 15516. Target Synonyms: HSP90-BETA; HSP90B; HSPC2; HSPCB; HSP84; Heat Shock 90kD Protein 1,Beta; Heat shock 84 kDa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2700