Learn More
Biomatik Corporation Human Indoleamine-2, 3-Dioxygenase (IDO) ELISA Kit, 96 tests
Catalog No. 501502788
Encompass_Preferred
The Human Indoleamine-2, 3-Dioxygenase (IDO) ELISA Kit allows for quantitative determination indoleamine-2, 3-Dioxygenase in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.118 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P14902. Cross Activity: No significant cross-reactivity or interference between Indoleamine-2, 3-Dioxygenase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2788 Supplier Biomatik Corporation Supplier No. EKU04916
The Human Indoleamine-2, 3-Dioxygenase (IDO) ELISA Kit allows for quantitative determination indoleamine-2, 3-Dioxygenase in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.118 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P14902. Cross Activity: No significant cross-reactivity or interference between Indoleamine-2, 3-Dioxygenase and analogues was observed. Restriction: For Research Use Only.