Learn More
Biomatik Corporation Phosphocreatine (PCr) ELISA Kit
Catalog No. 502286898
Encompass_Preferred
Phosphocreatine (PCr) ELISA Kit for detecting PCr in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 123.5-10,000ng/ml. Sensitivity: 50.3ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: CP; Creatine Phosphate; Phosphorylcreatine; Creatine-P; Phosphagen; Fosfocreatine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6898 Supplier Biomatik Corporation Supplier No. EKU11336
Phosphocreatine (PCr) ELISA Kit for detecting PCr in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 123.5-10,000ng/ml. Sensitivity: 50.3ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: CP; Creatine Phosphate; Phosphorylcreatine; Creatine-P; Phosphagen; Fosfocreatine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.