Learn More
Biomatik Corporation Mouse Lymphotoxin Beta (LTb) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05707
Encompass_Preferred
Mouse Lymphotoxin Beta (LTb) ELISA Kit for detecting LTb in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P41155. Gene ID: 16994. Target Synonyms: LT-B; p33; TNFC; TNF-C; TNFSF3; Tumor necrosis factor ligand superfamily member 3; Tumor Necrosis Factor C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3162