Learn More
Biomatik Corporation Human Pleiotrophin (PTN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06716
Encompass_Preferred
The Human Pleiotrophin (PTN) ELISA Kit allows for quantitative determination of Pleiotrophin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2,000 pg/mL. Sensitivity: 11.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P21246. Cross Activity: No significant cross-reactivity or interference between Pleiotrophin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3557