Learn More
Biomatik Corporation Mouse Proopiomelanocortin (POMC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06842
Encompass_Preferred
Mouse Proopiomelanocortin (POMC) ELISA Kit for detecting POMC in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 48.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01193. Gene ID: 18976. Target Synonyms: POC; ProOMC; Pro-OMC; Pro Opiomelanocortin; Corticotropin-lipotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3615