Learn More
Biomatik Corporation Mouse Semaphorin 3A (SEMA3A) ELISA Kit, 96 tests
Catalog No. 501503774
Encompass_Preferred
Mouse Semaphorin 3A (SEMA3A) ELISA Kit for detecting SEMA3A in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08665. Gene ID: 20346. Target Synonyms: SEMAL; Hsema-I; Hsema-III; SEMA1; SEMAD; SEMAIII; SemD; Coll-1; Sema Domain,Immunoglobulin Domain (Ig) ,Transmembrane Domain (TM) and Short Cytoplasmic Domain 3A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3774 Supplier Biomatik Corporation Supplier No. EKU07258
Mouse Semaphorin 3A (SEMA3A) ELISA Kit for detecting SEMA3A in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08665. Gene ID: 20346. Target Synonyms: SEMAL; Hsema-I; Hsema-III; SEMA1; SEMAD; SEMAIII; SemD; Coll-1; Sema Domain,Immunoglobulin Domain (Ig) ,Transmembrane Domain (TM) and Short Cytoplasmic Domain 3A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.