Learn More
Biomatik Corporation Human Thioredoxin (Trx) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07613
Encompass_Preferred
Human Thioredoxin (Trx) ELISA Kit for detecting Trx in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.22ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10599. Gene ID: 7295. Target Synonyms: TXN; ADF; SASP; TRDX; TRX1; ATL-derived factor; Surface-associated sulphydryl protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3903