Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Ligand Superfamily, Member 14 (TNFSF14) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07967
Encompass_Preferred
Mouse Tumor Necrosis Factor Ligand Superfamily, Member 14 (TNFSF14) ELISA Kit for detecting TNFSF14 in serum, plasma, saliva, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QYH9. Gene ID: 50930. Target Synonyms: CD258; TR2; HVEML; LIGHT; LTg; Herpes virus entry mediator ligand; Herpesvirus entry mediator ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4059