Learn More
Biomatik Corporation Rat Tyrosine Hydroxylase (TH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08004
Encompass_Preferred
Rat Tyrosine Hydroxylase (TH) ELISA Kit for detecting TH in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04177. Gene ID: 25085. Target Synonyms: TYH; DYT5b; Tyrosine 3-Monooxygenase; Dystonia 14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4074