Learn More
Biomatik Corporation Rabbit Cortisol (Cor) ELISA Kit
Supplier:  Biomatik Corporation EKU11570
Encompass_Preferred
Rabbit Cortisol (Cor) ELISA Kit for detecting Cor in serum, plasma, urine, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 12.35-1,000ng/ml. Sensitivity: 4.84ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Hydrocortisone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6953