Learn More
Biomatik Corporation High Sensitive Ovalbumin (OVA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08280
Encompass_Preferred
The High Sensitive Ovalbumin (OVA) ELISA Kit allows for quantitative determination of Ovalbumin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: All species. Detection Range: 78-5000 pg/mL. Sensitivity: 30 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. Cross Activity: No significant cross-reactivity or interference between Ovalbumin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4183