Learn More
Biomatik Corporation Mouse Angiopoietin Like Protein 4 (ANGPTL4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02375
Encompass_Preferred
Mouse Angiopoietin Like Protein 4 (ANGPTL4) ELISA Kit for detecting ANGPTL4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z1P8. Gene ID: 57875. Target Synonyms: ARP4; FIAF; HFARP; NL2; PGAR; Pp1158; Fasting-Induced Adipose Factor; Hepatic Angiopoietin-Related Protein; Hepatic Fibrinogen/Angiopoietin-Related Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1795