Learn More
Biomatik Corporation Rabbit Collagen Type I (COL1) ELISA Kit
Supplier:  Biomatik Corporation EKU11673
Encompass_Preferred
Rabbit Collagen Type I (COL1) ELISA Kit for detecting COL1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G1T4A5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6983