Learn More
Biomatik Corporation Mouse Tight Junction Protein 1 (TJP1) ELISA Kit
Supplier:  Biomatik Corporation EKU11710
Encompass_Preferred
Mouse Tight Junction Protein 1 (TJP1) ELISA Kit for detecting TJP1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P39447. Gene ID: 21872. Target Synonyms: ZO1; Zona Occludens 1; Zona occludens protein 1; Zonula occludens protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6988