Learn More
Biomatik Corporation Human Sperm Adhesion Molecule 1 (SPAM1) ELISA Kit
Supplier:  Biomatik Corporation EKU11724
Encompass_Preferred
Human Sperm Adhesion Molecule 1 (SPAM1) ELISA Kit for detecting SPAM1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P38567. Gene ID: 6677. Target Synonyms: HYAL1; HYA1; HYAL3; HYAL5; PH-20; PH20; SPAG15; PH-20 Hyaluronidase; Zona Pellucida Binding. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6991