Learn More
Biomatik Corporation Human Peripherin (PRPH) ELISA Kit
Supplier:  Biomatik Corporation EKU12047
Encompass_Preferred
Human Peripherin (PRPH) ELISA Kit for detecting PRPH in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P41219. Gene ID: 5630. Target Synonyms: NEF4; Neurofilament 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7056