Learn More
Biomatik Corporation Human Fukutin Related Protein (FKRP) ELISA Kit
Supplier:  Biomatik Corporation EKU12049
Encompass_Preferred
Human Fukutin Related Protein (FKRP) ELISA Kit for detecting FKRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H9S5. Gene ID: 79147. Target Synonyms: LGMD2I; MDC1C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7058