Learn More
Biomatik Corporation Canine Inhibin Beta-B (Inhbb) Elisa Kit
Supplier:  Biomatik Corporation EKC31857
Encompass_Preferred
Canine Inhibin Beta-B (Inhbb) Elisa Kit for detecting Inhibin Beta-B (Inhbb) in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 5-1000pg/ml. Sensitivity: 2.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: F1PB74. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7232