Learn More
Biomatik Corporation Human Complement Factor H-Related Protein 3 (Cfhr3) Elisa Kit
Supplier:  Biomatik Corporation EKC33268
Encompass_Preferred
Human Complement Factor H-Related Protein 3 (Cfhr3) Elisa Kit for detecting Complement Factor H-Related 3 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000ng/ml. Sensitivity: 7.8ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q02985; CFHR3. Target Synonyms: CFHR3; CFHL3; FHR3Complement factor H-related protein 3; FHR-3; DOWN16; H factor-like protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7380