Learn More
Biomatik Corporation Human Complement Fragment 3D, C3D Elisa Kit
Supplier:  Biomatik Corporation EKC33276
Encompass_Preferred
Human Complement Fragment 3D, C3D Elisa Kit for detecting Complement Fragment 3D, C3D in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ug/ml. Sensitivity: 0.039ug/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7382