Learn More
Biomatik Corporation Mouse Cortisol ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC31387
Encompass_Preferred
Mouse Cortisol Elisa Kit for detecting Cortisol in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.049-200ng/ml. Sensitivity: 0.049ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-7445