Learn More
Biomatik Corporation Human Endothelial Nitric Oxide Synthase, Enos Elisa Kit
Supplier:  Biomatik Corporation EKC33569
Encompass_Preferred
Human Endothelial Nitric Oxide Synthase, Enos Elisa Kit for detecting Nitric Oxide Synthase 3 (Endothelial Cell) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.1-70IU/ml. Sensitivity: 0.27IU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P29474; NOS3. Target Synonyms: cNOS; Constitutive NOS; EC NOS; EC-NOS; ecNOS; Endothelial nitric oxidase synthase; Endothelial nitric oxide synthase; Endothelial nitric oxide synthase 3; Endothelial NOS; eNOS; Nitric oxide synthase 3 (endothelial cell); Nitric oxide synthase 3; Nitric oxide synthase 3 endothelial cell; Nitric oxide synthase endothelial; Nitric oxide synthase; endothelial; NOS 3; NOS III; NOS type III; NOS3; NOS3_HUMAN; NOSIII. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7403