Learn More
Biomatik Corporation Human fetal Fibronectin(fFN)ELISA Kit
Supplier:  Biomatik Corporation EKC33673
Encompass_Preferred
Sample Type: serum, plasma, urine, cell culture supernates. Species Reactivity: Homo sapiens (Human). Detection Range: 25 ng/mL-1600 ng/mL. Sensitivity: 6.25 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7412