Learn More
Biomatik Corporation Human Fetuin A Elisa Kit
Supplier:  Biomatik Corporation EKC33675
Encompass_Preferred
Human Fetuin A Elisa Kit for detecting Alpha-2-Hs-Glycoprotein in serum, plasma, cell culture supernates, tissue homogenates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000ng/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02765; AHSG. Target Synonyms: 59 kDa bone sialic acid-containing protein; A2HS; Aa2-066; AHS; Ahsg alpha-2-HS-glycoprotein; Ahsg; Alpha 2 HS Glycoprotein; Alpha 2 Z globulin; Alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; Asialofetuin; Ba alpha 2 glycoprotein; Ba-alpha-2-glycoprotein; BSP; Countertrypin; Fetua; FETUA_HUMAN; Fetuin; mouse; homolog of; Fetuin A; Fetuin-A; Glycoprotein PP63; HSGA; pp63. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7414