Learn More
Biomatik Corporation Human Fibroblast Growth Factor-23, Fgf-23 Elisa Kit
Supplier:  Biomatik Corporation EKC33708
Encompass_Preferred
Human Fibroblast Growth Factor-23, Fgf-23 Elisa Kit for detecting Fibroblast Growth Factor 23 in serum, cell culture supernates, urine, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9GZV9; FGF23. Target Synonyms: ADHR; FGF-23; Fgf23; FGF23_HUMAN; FGFN; Fibroblast growth factor 23; Fibroblast growth factor 23 C-terminal peptide; Fibroblast growth factor 23 precursor; HPDR2; HYPF; Phosphatonin; PHPTC; Tumor derived hypophosphatemia inducing factor; Tumor-derived hypophosphatemia-inducing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7418