Learn More
Biomatik Corporation Human Gastrin-Releasing Peptide, Grp Elisa Kit
Supplier:  Biomatik Corporation EKC33803
Encompass_Preferred
Human Gastrin-Releasing Peptide, Grp Elisa Kit for detecting Gastrin-Releasing Peptide in serum, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.312ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07492; GRP. Target Synonyms: BN; Bombesin; GRP; GRP-10; GRP_HUMAN; GRP10; Neuromedin-C; NeuromedinC; Pre progastrin releasing peptide; PreproGRP; ProGRP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7420