Learn More
Biomatik Corporation Human Glutamic Acid Decarboxylase 65, Gad65 Elisa Kit
Supplier:  Biomatik Corporation EKC33846
Encompass_Preferred
Human Glutamic Acid Decarboxylase 65, Gad65 Elisa Kit for detecting Glutamate Decarboxylase 2 (Pancreatic Islets And Brain, 65Kda) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q05329; GAD2. Target Synonyms: 65 kDa glutamic acid decarboxylase; DCE 2; DCE2; DCE2_HUMAN; GAD 2; GAD 65; GAD-2; GAD-65; Gad2; Glutamate decarboxylase 2 (pancreas); Glutamate Decarboxylase 2 (pancreatic islets and brain 65kDa); Glutamate decarboxylase 2; Glutamate Decarboxylase 65; Glutamate decarboxylase 65 kDa isoform; Glutamic Acid Decarboxylase 2; Glutamic Acid Decarboxylase 65; MGC161605; MGC161607. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7425