Learn More
Biomatik Corporation Human Interleukin 21, Il-21 Elisa Kit
Supplier:  Biomatik Corporation EKC34234
Encompass_Preferred
Human Interleukin 21, Il-21 Elisa Kit for detecting Interleukin 21 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9HBE4; IL21. Target Synonyms: CVID11; IL 21; IL-21; Il21; IL21_HUMAN; Interleukin 21; Interleukin-21; interleukin-21 isoform; Interleukin21; OTTHUMP00000164088; Za11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7454