Learn More
Biomatik Corporation Human Interleukin 3, Il-3 Elisa Kit
Supplier:  Biomatik Corporation EKC34244
Encompass_Preferred
Human Interleukin 3, Il-3 Elisa Kit for detecting Interleukin 3 (Colony-Stimulating Factor, Multiple) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P08700; IL3. Target Synonyms: Colony stimulating factor multiple; Hematopoietic growth factor; IL 3; IL-3; IL3; IL3_HUMAN; Interleukin 3 (colony stimulating factor; multiple); Interleukin 3; Interleukin-3; Mast cell growth factor; MCGF; MGC79398; MGC79399; Multi CSF; Multilineage colony stimulating factor; Multipotential colony stimulating factor; Multipotential colony-stimulating factor; OTTHUMP00000065963; P cell stimulating factor; P-cell-stimulating factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7456