Learn More
Biomatik Corporation Human Intestinal Fatty Acid Binding Protein, Ifabp Elisa Kit
Supplier:  Biomatik Corporation EKC34306
Encompass_Preferred
Human Intestinal Fatty Acid Binding Protein, Ifabp Elisa Kit for detecting Fatty Acid Binding Protein 2, Intestinal in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 2.5-160ng/ml. Sensitivity: 0.625ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P12104; FABP2. Target Synonyms: FABP 2; FABP2; FABPI; FABPI_HUMAN; Fatty acid binding protein 2 intestinal; Fatty acid binding protein; Fatty acid binding protein intestinal; Fatty acid-binding protein 2; Fatty acid-binding protein; I FABP; I-FABP; IFABP; intestinal; intestinal FABP; Intestinal fatty acid binding protein 2; Intestinal-type fatty acid-binding protein; MGC133132; OTTHUMP00000163925. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7461